Learn More
Selleck Chemical LLC Celecoxib 100mg 169590-42-5 SC 58635
SDP
Catalog No. 507847
Encompass_Preferred
Celecoxib (SC 58635) is a selective COX-2 inhibitor with IC50 of 40 nM in Sf9 cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-784-7 Supplier Selleck Chemical LLC Supplier No. S1261100MG
Celecoxib (SC 58635) is a selective COX-2 inhibitor with IC50 of 40 nM in Sf9 cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.