Learn More
Regis Technologies Inc CELERIS ARGININE 100MMX3MM CLN
SDP
Catalog No. NC2267532
Encompass
Celeris Arginine, 100 mm x 3 mm, 5 μm, Analytical Column
This item is not returnable. View return policy
Catalog No. NC2267532 Supplier Regis Technologies Inc Supplier No. 1790913300
Celeris Arginine, 100 mm x 3 mm, 5 μm, Analytical Column