Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Celestial Globe with Meridian Ring
GREENER_CHOICESCI_ED

Catalog No. S816747 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
6+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S816747 6+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S816747 Supplier Supertek™ Supplier No. B19018TSP
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Create your own planetarium and study the stars with this Celestial Globe with Meridian Ring. This globe is removable from the cradle and makes it easy to demonstrate Earth's apparent relationship to the stars, planets, and galaxies.

A perfect medium to define star activities to students, this unique 12 in. Basic Celestial Globe with 3.3 in. internal world globe is adjustable and made of superior quality polymers. The sun may be set by adjusting the sun knob located near the North Pole. The globe can be set to reflect constellation position according to time and place to show the actual positions of the stars, and the outer surface can be marked and cleaned. The meridian may be tilted to adjust the globe for latitude, while the horizon ring indicates the actual horizon in relation to the point of observation on Earth.

This hands-on globe experience makes it more practical and fun to examine the universe. Comes with base and teaching aids.

  • Globe-in-globe configuration
  • Globe may be removed from cradle for closer view
  • Mounted in full horizon and meridian base for stability
  • Markable outer surface
  • Easily washable
  • Milky Way depicted in white
  • Stars depicted in yellow
  • Comes with 19 page activity book and 8 page study guide

Specifications

Grade 6+
Format Manual
Product Type Celestial Globe
Includes Globe, Extensive Teaching Aids
No. of Pages 19 (Activity Book), 8 (Study Guide)
Languages English
For Use With (Application) Teaching of Stars, Planets and Galaxies
Subject Earth and Space
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.