Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Celestine Blue, pure

Catalog No. AC195560250
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC195560250 25 g
AC195560100 10 g
AC195560500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC195560250 Supplier Thermo Scientific Chemicals Supplier No. 195560250

CAS: 1562-90-9 | C17H18ClN3O4 | 363.798 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1562-90-9
Molecular Formula C17H18ClN3O4
Molecular Weight (g/mol) 363.798
MDL Number MFCD00011927
InChI Key OJAMERVLRXYMPD-UHFFFAOYSA-N
PubChem CID 54684697
IUPAC Name 7-(diethylamino)-4-hydroxy-3-oxophenoxazin-10-ium-1-carboxamide;chloride
SMILES CCN(CC)C1=CC2=C(C=C1)[NH+]=C3C(=CC(=O)C(=C3O2)O)C(=O)N.[Cl-]

Specifications

Merck Index 15, 1961
Infrared Spectrum Authentic
Lambda Max 638 to 648nm
Formula Weight 363.79
Grade Extra Pure
Specific Extinction 700 min. (in water)
UV Spectrometry Complies
Color Black to Brown
Quantity 25 g
Physical Form Powder
Chemical Name or Material Celestine blue
Show More Show Less
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.