Learn More
Medchemexpress LLC Celgosivir (hydrochloride) | 141117-12-6 | 98.0% | 295.76 | 10 MM * 1 ML
SDP
Catalog No. 5000391489
Encompass_Preferred
Celgosivir hydrochloride is an α-glucosidase I inhibitor that inhibits bovine viral diarrhoea virus (BVDV) with an IC50 of 1.27 μM in an in vitro assay.
  • Acts as an α-glucosidase I inhibitor.
  • Exhibits antiviral activity.
  • Potent against HIV-1.
  • Active against BVDV.
  • Active against DENV2.
  • Active against DENV1, 3, and 4.
  • Offers in vivo protection.
  • Provides dose and schedule-dependent protection.
  • Undergoes rapid metabolism.
  • Appears as a solid, white to gray color.
  • Ships at room temperature in continental US (may vary elsewhere). Store at 4°C, sealed, away from moisture. In solvent, store at -80°C for 6 months or -20°C for 1 month (sealed, away from moisture).
Catalog No. 50-003-91489 Supplier Medchemexpress LLC Supplier No. HY16134A10MM/1ML
Celgosivir hydrochloride is an α-glucosidase I inhibitor that inhibits bovine viral diarrhoea virus (BVDV) with an IC50 of 1.27 μM in an in vitro assay.
  • Acts as an α-glucosidase I inhibitor.
  • Exhibits antiviral activity.
  • Potent against HIV-1.
  • Active against BVDV.
  • Active against DENV2.
  • Active against DENV1, 3, and 4.
  • Offers in vivo protection.
  • Provides dose and schedule-dependent protection.
  • Undergoes rapid metabolism.
  • Appears as a solid, white to gray color.
  • Ships at room temperature in continental US (may vary elsewhere). Store at 4°C, sealed, away from moisture. In solvent, store at -80°C for 6 months or -20°C for 1 month (sealed, away from moisture).