Learn More
Selleck Chemical LLC Celiprolol hydrochloride
SDP
Catalog No. 5000003909
Encompass_Preferred
Celiprolol is a 1-adrenoceptor antagonist with partial 2 agonist activity
Catalog No. 50-000-03909 Supplier Selleck Chemical LLC Supplier No. S592310MG
Celiprolol is a 1-adrenoceptor antagonist with partial 2 agonist activity