Learn More
Cayman Chemical Celiprolol (hydrochloride), Purity: ≥98%, CAS: 57470-78-7, 10 mg
SDP
Catalog No. 502965445
Encompass_Preferred
A β1 adrenergic receptor antagonist and a β2 adrenergic receptor partial agonist that relaxes human arteries and veins with ED50 values of 40-50 µM in vitro; reduces myocardial infarct size and increases NO production in a rabbit model of myocardial ischemia at 1-10 mg/kg/h
Catalog No. 50-296-5445 Supplier Cayman Chemical Supplier No. 1671510 MG
A β1 adrenergic receptor antagonist and a β2 adrenergic receptor partial agonist that relaxes human arteries and veins with ED50 values of 40-50 µM in vitro; reduces myocardial infarct size and increases NO production in a rabbit model of myocardial ischemia at 1-10 mg/kg/h