Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Celite™ 545, MilliporeSigma™

Catalog No. M1026931000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 g
1 kg
10 kg
Packaging:
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1026931000 1 kg Plastic Bottle
M1026930250 250 g Plastic Bottle
M1026939010 10 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1026931000 Supplier MilliporeSigma Supplier No. 1.02693.1000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recommended Storage

Storage temperature: No restrictions

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68855-54-9
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.988
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium;carbonate
SMILES C(=O)([O-])[O-].[Na+].[Na+]

Specifications

pH 10 (100 g/L, H2O, 20°C)
Quantity 1 kg
Particle Size Particle size (0.1mm): > 60%
Packaging Plastic Bottle
Solubility Information <0.001g/L
EINECSNumber 272-489-0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.