Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Celite 545 Filter Aid (Powder), Fisher Chemical™

Catalog No. C21225LB Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
25 lb.
50 lb.
Packaging:
Fiber Drum
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
C212-25LB 25 lb. Fiber Drum
C212-500 500 g Poly Bottle
C212-50LB 50 lb. Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C212-25LB
Add to Cart

Not acid-washed

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68855-54-9
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.988
MDL Number MFCD00132803
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym Diatomaceous Earth, Kieselguhr
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium;carbonate
SMILES C(=O)([O-])[O-].[Na+].[Na+]

Specifications

Physical Form Solid
Density 2.3 g/cm3
Quantity 25 lb.
Color White
pH 9 to 10.5
Packaging Fiber Drum
Chemical Name or Material Celite™ 545 Filter Aid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.