Learn More
Perkin Elmer US LLC NIR Quartz SUPRASIL 300 Rectangular Macro Cell with Lid - 10 mm, Qty. 2
Catalog No. 509052046
Encompass_Preferred
The NIR Quartz SUPRASIL 300 Macro Cell has a volume of 3.5 mL, a path length of 10 mm, a PTFE lid, and outside dimensions of 45 mm (H) x 12.5 mm (W) x 12.5 mm (D). Qty. 2
Catalog No. 50-905-2046 Supplier Perkin Elmer US LLC Supplier No. B0631015
The NIR Quartz SUPRASIL 300 Macro Cell has a volume of 3.5 mL, a path length of 10 mm, a PTFE lid, and outside dimensions of 45 mm (H) x 12.5 mm (W) x 12.5 mm (D). Qty. 2