Learn More
Perkin Elmer US LLC UV Quartz SUPRASIL Rectangular Macro Cell with PTFE Stopper - 1 mm, Qty. 2
Catalog No. 509052051
Encompass_Preferred
The UV Quartz SUPRASIL Macro Cell has a 10 mm path length, a cell volume of 3.5 mL, and a PTFE lid. Its outside dimensions are 45 mm (H) x 12.5 mm (W) x 12.5 mm (D). Qty. 2
Catalog No. 50-905-2051 Supplier Perkin Elmer US LLC Supplier No. B0631025
The UV Quartz SUPRASIL Macro Cell has a 10 mm path length, a cell volume of 3.5 mL, and a PTFE lid. Its outside dimensions are 45 mm (H) x 12.5 mm (W) x 12.5 mm (D). Qty. 2