Learn More
Starna Cells Inc CELL 1Q10
SDP
Catalog No. NC9469798
Encompass_Preferred
Standard Rectangular Cells, Path Length: 10mm, Exterior Size: 12.5L x 12.5W x 45mmH, Interior Size: 10W x 10mm long, Nominal Volume: 3, 500 mL
Catalog No. NC9469798 Supplier Starna Cells Inc Supplier No. 1Q10
Standard Rectangular Cells, Path Length: 10mm, Exterior Size: 12.5L x 12.5W x 45mmH, Interior Size: 10W x 10mm long, Nominal Volume: 3, 500 mL