Learn More
Quality Biological Inc Cell Scraper, 25 cm Handle, 20mm Lifter Blade, Sterile, 100/CS
SDP
Catalog No. 50147433
Encompass_Preferred
Cell Scraper, 25 cm Handle, 20mm Lifter Blade, Sterile
Packaged Individually Wrapped in Bag, 100/CS
• Cell Scrapers are designed to gently harvest cells from tissue culture flasks, dishes, or bottles
• Soft, pliable blade won’t damage cells or scratch surfaces
• Lifter blade is positioned perpendicular to the handle for use with a raking motion
Catalog No. 50-147-433 Supplier Quality Biological Inc Supplier No. P886229311
Cell Scraper, 25 cm Handle, 20mm Lifter Blade, Sterile
Packaged Individually Wrapped in Bag, 100/CS
• Cell Scrapers are designed to gently harvest cells from tissue culture flasks, dishes, or bottles
• Soft, pliable blade won’t damage cells or scratch surfaces
• Lifter blade is positioned perpendicular to the handle for use with a raking motion