Learn More
Quality Biological Inc Cell Scraper, 25 cm Handle, 20mm Scraper Blade, Sterile, 100/CS
SDP
Catalog No. 50147432
Encompass_Preferred
Cell Scraper, 25 cm Handle, 20mm Scraper Blade, Sterile
Packaged Individually Wrapped in Bag, 100/CS
• Cell Scrapers are designed to gently harvest cells from tissue culture flasks, dishes, or bottles
• Soft, pliable blade won’t damage cells or scratch surfaces
• Scraper blade is positioned parallel to the handle for windshield wiper like scraping
Catalog No. 50-147-432 Supplier Quality Biological Inc Supplier No. P886229310
Cell Scraper, 25 cm Handle, 20mm Scraper Blade, Sterile
Packaged Individually Wrapped in Bag, 100/CS
• Cell Scrapers are designed to gently harvest cells from tissue culture flasks, dishes, or bottles
• Soft, pliable blade won’t damage cells or scratch surfaces
• Scraper blade is positioned parallel to the handle for windshield wiper like scraping