Learn More
Sorbent Technologies Inc Cellulose 300 TLC Plates, aluminum backed, 100um, 4x8cm, 50pk
SDP
Catalog No. 501138508
Encompass_Preferred
Cellulose 300 aluminum backed TLC plates. The coating thickness is 100um and dimensions are 4x8cm. There are 50 plates per pack. Applications include: partition chromatography of polar substances such as amino acids, carboxylic acids or carbohydrates. Cellulose is especially used for partition chromatographic separations of polar substances such as amino acids and other carboxylic acids or carbohydrates. Compared to paper chromatography (PC) thin layer chromatography, cellulose offers shorter migration times and more concentrated substance spots resulting in a higher detection sensitivity. TLC plates (CEL 300) are coated with native, fibrous cellulose.
Catalog No. 50-113-8508 Supplier Sorbent Technologies Inc Supplier No. 0332087
Cellulose 300 aluminum backed TLC plates. The coating thickness is 100um and dimensions are 4x8cm. There are 50 plates per pack. Applications include: partition chromatography of polar substances such as amino acids, carboxylic acids or carbohydrates. Cellulose is especially used for partition chromatographic separations of polar substances such as amino acids and other carboxylic acids or carbohydrates. Compared to paper chromatography (PC) thin layer chromatography, cellulose offers shorter migration times and more concentrated substance spots resulting in a higher detection sensitivity. TLC plates (CEL 300) are coated with native, fibrous cellulose.