Learn More
Sorbent Technologies Inc Cellulose 300 TLC Plates, polyester backed, 100um, 5x20cm, 50/pk
SDP
Catalog No. 501138502
Encompass_Preferred
Cellulose 300 polyester backed TLC plates. The coating thickness is 100um and dimensions are 5x20cm. There are 50 plates per pack. Applications include: partition chromatography of polar substances such as amino acids, carboxylic acids or carbohydrates. Cellulose is especially used for partition chromatographic separations of polar substances such as amino acids and other carboxylic acids or carbohydrates. Compared to paper chromatography (PC) thin layer chromatography, cellulose offers shorter migration times and more concentrated substance spots resulting in a higher detection sensitivity. TLC plates (CEL 300) are coated with native, fibrous cellulose.
Catalog No. 50-113-8502 Supplier Sorbent Technologies Inc Supplier No. 0322047
Cellulose 300 polyester backed TLC plates. The coating thickness is 100um and dimensions are 5x20cm. There are 50 plates per pack. Applications include: partition chromatography of polar substances such as amino acids, carboxylic acids or carbohydrates. Cellulose is especially used for partition chromatographic separations of polar substances such as amino acids and other carboxylic acids or carbohydrates. Compared to paper chromatography (PC) thin layer chromatography, cellulose offers shorter migration times and more concentrated substance spots resulting in a higher detection sensitivity. TLC plates (CEL 300) are coated with native, fibrous cellulose.