Learn More
Sorbent Technologies Inc Cellulose 300 TLC Plates, w/UV254, glass backed, 250um, 20x20cm, 25pk
SDP
Catalog No. 501138497
Encompass_Preferred
Cellulose 300 with UV254, glass backed TLC plates. The coating thickness is 250um and dimensions are 20x20cm. There are 25 plates per pack. Applications include: partition chromatography of polar substances such as amino acids, carboxylic acids or carbohydrates. Cellulose is especially used for partition chromatographic separations of polar substances such as amino acids and other carboxylic acids or carbohydrates. Compared to paper chromatography (PC) thin layer chromatography, cellulose offers shorter migration times and more concentrated substance spots resulting in a higher detection sensitivity. TLC plates (CEL 300) are coated with native, fibrous cellulose.
Catalog No. 50-113-8497 Supplier Sorbent Technologies Inc Supplier No. 0315126
Cellulose 300 with UV254, glass backed TLC plates. The coating thickness is 250um and dimensions are 20x20cm. There are 25 plates per pack. Applications include: partition chromatography of polar substances such as amino acids, carboxylic acids or carbohydrates. Cellulose is especially used for partition chromatographic separations of polar substances such as amino acids and other carboxylic acids or carbohydrates. Compared to paper chromatography (PC) thin layer chromatography, cellulose offers shorter migration times and more concentrated substance spots resulting in a higher detection sensitivity. TLC plates (CEL 300) are coated with native, fibrous cellulose.