Learn More
Sorbent Technologies Inc Cellulose 400 TLC Plates, w/UV254, glass backed, 100um, 20x20cm, 25/pk
SDP
Catalog No. 501138513
Encompass_Preferred
Cellulose 400 (microcrystalline cellulose AVICEL) with UV254, glass backed TLC plates. The coating thickness is 100um and dimensions are 20x20cm. There are 25 plates per pack. Applications include: carboxylic acids, lower alcohols, urea and purine derivatives. Cellulose 400 TLC Plates are coated with microcrystalline cellulose AVICEL. This type of cellulose is prepared by hydrolysis of high purity cellulose with hydrochloric acid. Depending on the starting cellulose and the following clean-up, one obtains cellulose crystallites with a mean degree of polymerization between 40 and 200. The microcrystalline structure of this cellulose can be confirmed by X-ray investigations.
Catalog No. 50-113-8513 Supplier Sorbent Technologies Inc Supplier No. 0412126
Cellulose 400 (microcrystalline cellulose AVICEL) with UV254, glass backed TLC plates. The coating thickness is 100um and dimensions are 20x20cm. There are 25 plates per pack. Applications include: carboxylic acids, lower alcohols, urea and purine derivatives. Cellulose 400 TLC Plates are coated with microcrystalline cellulose AVICEL. This type of cellulose is prepared by hydrolysis of high purity cellulose with hydrochloric acid. Depending on the starting cellulose and the following clean-up, one obtains cellulose crystallites with a mean degree of polymerization between 40 and 200. The microcrystalline structure of this cellulose can be confirmed by X-ray investigations.