Learn More
Eurofins Discoverx Cell Activation Cocktail, 100 µL. (92-1334)
Catalog No. 501999844
Encompass
The Cell Activation Cocktail is a pre-mixed reagent that includes PMA (phorbol 12-myristate-13-acetate) and ionomycin. The mixture does not include Brefeldin A. This reagent is used for T cell activation, and increases cell surface expression of certain cell surface markers (e.g. CD69). Provided as 100 μL of 500x premixed solution. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9844 Supplier Eurofins Discoverx Supplier No. 921334
The Cell Activation Cocktail is a pre-mixed reagent that includes PMA (phorbol 12-myristate-13-acetate) and ionomycin. The mixture does not include Brefeldin A. This reagent is used for T cell activation, and increases cell surface expression of certain cell surface markers (e.g. CD69). Provided as 100 μL of 500x premixed solution. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.