Learn More
Cell Biolabs Inc CELL ADHESION ASSAY- 48 ASSAYS
Catalog No. NC2391632
Encompass_Preferred
Full quantitation of cell adhesion with no manual cell countingFluorometric or colorimetric detectionPlates precoated with a uniform substrate layer of a single ECM protein in each row: Collagen I, Collagen IV, Fibrinogen, Fibronectin, and Laminin.Our CytoSelect™ 48-Well Cell Adhesion Assays provide a fully quantitative method for the evaluation of cell adhesion. The 48-well plate is precoated with your choice of single ECM protein in each of the first 5 rows, with the last row provided as a negative control. Just follow a simple protocol:Cells are seeded onto the substrate.Adherent cells attach, while non-adherent cells are washed away.Adherent cells are quantified on a standard plate reader or fluorometer.
Catalog No. NC2391632 Supplier Cell Biolabs Inc Supplier No. CBA0705
Full quantitation of cell adhesion with no manual cell countingFluorometric or colorimetric detectionPlates precoated with a uniform substrate layer of a single ECM protein in each row: Collagen I, Collagen IV, Fibrinogen, Fibronectin, and Laminin.Our CytoSelect™ 48-Well Cell Adhesion Assays provide a fully quantitative method for the evaluation of cell adhesion. The 48-well plate is precoated with your choice of single ECM protein in each of the first 5 rows, with the last row provided as a negative control. Just follow a simple protocol:Cells are seeded onto the substrate.Adherent cells attach, while non-adherent cells are washed away.Adherent cells are quantified on a standard plate reader or fluorometer.