Learn More
Eurofins Discoverx PathHunter Human HTR6 (5-HT6) β-Arrestin Cell-Based Assay Kit (DLD1)
Catalog No. 5000455882
Encompass
The PathHunter Human HTR6 (5-HT6) β-Arrestin Stable Cell Line Assay (DLD1) contains a stable clonal cell line used to measure HTR6 (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55882 Supplier Eurofins Discoverx Supplier No. 7930294C13
The PathHunter Human HTR6 (5-HT6) β-Arrestin Stable Cell Line Assay (DLD1) contains a stable clonal cell line used to measure HTR6 (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.