Learn More
Eurofins Discoverx PathHunter Human PTGER4 β-Arrestin Cell-Based Assay Kit (HEK 293)
Catalog No. 5000456000
Encompass
The PathHunter Human PTGER4 β-Arrestin Stable Cell Line Assay (HEK 293) contains a stable clonal cell line used to measure PTGER4 (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56000 Supplier Eurofins Discoverx Supplier No. 7930304C1
Add to Cart
Add to Cart
The PathHunter Human PTGER4 β-Arrestin Stable Cell Line Assay (HEK 293) contains a stable clonal cell line used to measure PTGER4 (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.