Learn More
Eurofins Discoverx PathHunter Human PTGIR β-Arrestin Cell-Based Assay Kit (HEK 293)
Catalog No. 5000456178
Encompass
The PathHunter Human PTGIR β-Arrestin Stable Cell Line Assay (HEK 293) contains a stable clonal cell line used to measure PTGIR (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56178 Supplier Eurofins Discoverx Supplier No. 7930305C1
Add to Cart
Add to Cart
The PathHunter Human PTGIR β-Arrestin Stable Cell Line Assay (HEK 293) contains a stable clonal cell line used to measure PTGIR (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.