Learn More
Eurofins Discoverx PathHunter Human TACR1 β-Arrestin Cell-Based Assay Kit (CHO-K1)
Catalog No. 5000455997
Encompass
The PathHunter Human TACR1 β-Arrestin Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure TACR1 (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55997 Supplier Eurofins Discoverx Supplier No. 7930344C2
The PathHunter Human TACR1 β-Arrestin Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure TACR1 (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.