Learn More
Eurofins Discoverx SPRINTer™ Human BRD4 Targeted Protein Degradation Cell-Based Assay Kit (K562)
Catalog No. 5000455546
Encompass
The SPRINTer™ Human BRD4 Targeted Protein Degradation Stable Cell Line Assay (K562) contains a stable clonal cell line used for monitoring drug-induced protein turnover of BRD4, supporting TPD studies. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55546 Supplier Eurofins Discoverx Supplier No. 7911002C042
The SPRINTer™ Human BRD4 Targeted Protein Degradation Stable Cell Line Assay (K562) contains a stable clonal cell line used for monitoring drug-induced protein turnover of BRD4, supporting TPD studies. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.