Learn More
Eurofins Discoverx SPRINTer™ Human IKZF1 Targeted Protein Degradation Cell-Based Assay Kit (K562)
Catalog No. 5000455642
Encompass
The SPRINTer™ Human IKZF1 Targeted Protein Degradation Stable Cell Line Assay (K562) contains a stable clonal cell line used for monitoring drug-induced protein turnover of IKZF1, supporting TPD studies. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55642 Supplier Eurofins Discoverx Supplier No. 7911007C042
The SPRINTer™ Human IKZF1 Targeted Protein Degradation Stable Cell Line Assay (K562) contains a stable clonal cell line used for monitoring drug-induced protein turnover of IKZF1, supporting TPD studies. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.