Learn More
Eurofins Discoverx SPRINTer™ Human CDKN1A (p21) Targeted Protein Degradation Cell-Based Assay Kit (HCT-116)
Catalog No. 5000456082
Encompass
The SPRINTer™ Human CDKN1A (p21) Targeted Protein Degradation Stable Cell Line Assay (HCT-116) contains a stable clonal cell line used for monitoring drug-induced protein turnover of CDKN1A (p21), supporting TPD studies. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56082 Supplier Eurofins Discoverx Supplier No. 7911006C047
The SPRINTer™ Human CDKN1A (p21) Targeted Protein Degradation Stable Cell Line Assay (HCT-116) contains a stable clonal cell line used for monitoring drug-induced protein turnover of CDKN1A (p21), supporting TPD studies. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.