Learn More
Eurofins Discoverx SPRINTer™ Human c-Myc Targeted Protein Degradation Cell-Based Assay Kit (HCT-116)
Catalog No. 5000456213
Encompass
The SPRINTer™ Human c-Myc Targeted Protein Degradation Stable Cell Line Assay (HCT-116) contains a stable clonal cell line used for monitoring drug-induced protein turnover of c-Myc, supporting TPD studies. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56213 Supplier Eurofins Discoverx Supplier No. 7911003C047
The SPRINTer™ Human c-Myc Targeted Protein Degradation Stable Cell Line Assay (HCT-116) contains a stable clonal cell line used for monitoring drug-induced protein turnover of c-Myc, supporting TPD studies. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.