Learn More
Eurofins Discoverx SPRINTer™ Human BRAF(V600E) Targeted Protein Degradation Cell-Based Assay Kit (A375)
Catalog No. 5000456214
Encompass
The SPRINTer™ Human BRAF(V600E) Targeted Protein Degradation Stable Cell Line Assay (A375) contains a stable clonal cell line used for monitoring drug-induced protein turnover of BRAF(V600E), supporting TPD studies. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56214 Supplier Eurofins Discoverx Supplier No. 7911008C057
Add to Cart
Add to Cart
The SPRINTer™ Human BRAF(V600E) Targeted Protein Degradation Stable Cell Line Assay (A375) contains a stable clonal cell line used for monitoring drug-induced protein turnover of BRAF(V600E), supporting TPD studies. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.