Learn More
Eurofins Discoverx PathHunter Human GPR119 β-Arrestin Cell-Based Assay Kit (U2OS)
Catalog No. 5000456172
Encompass
The PathHunter Human GPR119 β-Arrestin Stable Cell Line Assay (U2OS) contains a stable clonal cell line used to measure GPR119 (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56172 Supplier Eurofins Discoverx Supplier No. 7930356C3
The PathHunter Human GPR119 β-Arrestin Stable Cell Line Assay (U2OS) contains a stable clonal cell line used to measure GPR119 (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.