Learn More
Eurofins Discoverx PathHunter Human GPR20 β-Arrestin Cell-Based Assay Kit (HEK 293)
Catalog No. 5000455652
Encompass
The PathHunter Human GPR20 β-Arrestin Stable Cell Line Assay (HEK 293) contains a stable clonal cell line used to measure GPR20 (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55652 Supplier Eurofins Discoverx Supplier No. 7930405C1
The PathHunter Human GPR20 β-Arrestin Stable Cell Line Assay (HEK 293) contains a stable clonal cell line used to measure GPR20 (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.