Learn More
Eurofins Discoverx PathHunter Human FXR Nuclear Receptor Protein Interaction Cell-Based Assay Kit (CHO-K1)
Catalog No. 5000455551
Encompass
The PathHunter Human FXR Nuclear Receptor Stable Cell Line Protein Interaction Assay (CHO-K1) contains a stable clonal cell line used to measure activated FXR nuclear hormone receptor interaction with SRCPs. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55551 Supplier Eurofins Discoverx Supplier No. 7930443C2
The PathHunter Human FXR Nuclear Receptor Stable Cell Line Protein Interaction Assay (CHO-K1) contains a stable clonal cell line used to measure activated FXR nuclear hormone receptor interaction with SRCPs. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.