Learn More
Eurofins Discoverx PathHunter Human RXRα Nuclear Receptor Protein Interaction Cell-Based Assay Kit (U2OS)
Catalog No. 5000455825
Encompass
The PathHunter Human RXRα Nuclear Receptor Stable Cell Line Protein Interaction Assay (U2OS) contains a stable clonal cell line used to measure activated RXRα nuclear hormone receptor interaction with SRCPs. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55825 Supplier Eurofins Discoverx Supplier No. 7930439C3
The PathHunter Human RXRα Nuclear Receptor Stable Cell Line Protein Interaction Assay (U2OS) contains a stable clonal cell line used to measure activated RXRα nuclear hormone receptor interaction with SRCPs. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.