Learn More
Eurofins Discoverx PathHunter Human MR Nuclear Translocation Cell-Based Assay Kit (CHO-K1)
Catalog No. 5000455554
Encompass
The PathHunter Human MR Stable Cell Line Nuclear Translocation Assay (CHO-K1) contains a stable clonal cell line used to measure MR nuclear hormone receptor activation via nuclear translocation. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55554 Supplier Eurofins Discoverx Supplier No. 7930451C2
Add to Cart
Add to Cart
The PathHunter Human MR Stable Cell Line Nuclear Translocation Assay (CHO-K1) contains a stable clonal cell line used to measure MR nuclear hormone receptor activation via nuclear translocation. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.