Learn More
Eurofins Discoverx SPRINTer™ Human KRAS(G12C) Targeted Protein Degradation Cell-Based Assay Kit (A549)
Catalog No. 5000456084
Encompass
The SPRINTer™ Human KRAS(G12C) Targeted Protein Degradation Stable Cell Line Assay (A549) contains a stable clonal cell line used for monitoring drug-induced protein turnover of KRAS(G12C), supporting TPD studies. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56084 Supplier Eurofins Discoverx Supplier No. 7911009C15
The SPRINTer™ Human KRAS(G12C) Targeted Protein Degradation Stable Cell Line Assay (A549) contains a stable clonal cell line used for monitoring drug-induced protein turnover of KRAS(G12C), supporting TPD studies. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.