Learn More
Eurofins Discoverx PathHunter Human AR Nuclear Translocation Cell-Based Assay Kit (HEK 293)
Catalog No. 5000456207
Encompass
The PathHunter Human AR Stable Cell Line Nuclear Translocation Assay (HEK 293) contains a stable clonal cell line used to measure AR nuclear hormone receptor activation via nuclear translocation. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56207 Supplier Eurofins Discoverx Supplier No. 7930177C1
The PathHunter Human AR Stable Cell Line Nuclear Translocation Assay (HEK 293) contains a stable clonal cell line used to measure AR nuclear hormone receptor activation via nuclear translocation. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.