Learn More
Eurofins Discoverx PathHunter Human Heregulin beta 1 (ErbB4) Functional Kinase Cell-Based Assay Kit (U2OS)
Catalog No. 5000455587
Encompass
The PathHunter Human Heregulin beta 1 (ErbB4) Stable Cell Line Functional Kinase Assay (U2OS) measures ErbB4 receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55587 Supplier Eurofins Discoverx Supplier No. 7930465C3
The PathHunter Human Heregulin beta 1 (ErbB4) Stable Cell Line Functional Kinase Assay (U2OS) measures ErbB4 receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.