Learn More
Eurofins Discoverx PathHunter Human PR beta Nuclear Translocation Cell-Based Assay Kit (U2OS)
Catalog No. 5000455496
Encompass
The PathHunter Human PR beta Stable Cell Line Nuclear Translocation Assay (U2OS) contains a stable clonal cell line used to measure PRβ nuclear hormone receptor activation via nuclear translocation. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55496 Supplier Eurofins Discoverx Supplier No. 7930484C3
The PathHunter Human PR beta Stable Cell Line Nuclear Translocation Assay (U2OS) contains a stable clonal cell line used to measure PRβ nuclear hormone receptor activation via nuclear translocation. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.