Learn More
Eurofins Discoverx PathHunter Human PDGFRb Functional Kinase Cell-Based Assay Kit (U2OS)
Catalog No. 5000456003
Encompass
The PathHunter Human PDGFRb Stable Cell Line Functional Kinase Assay (U2OS) measures PDGFRb receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56003 Supplier Eurofins Discoverx Supplier No. 7930493C3
Add to Cart
Add to Cart
The PathHunter Human PDGFRb Stable Cell Line Functional Kinase Assay (U2OS) measures PDGFRb receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.