Learn More
Eurofins Discoverx PathHunter Human IGF1R Functional Kinase Cell-Based Assay Kit (HEK 293)
Catalog No. 5000456179
Encompass
The PathHunter Human IGF1R Stable Cell Line Functional Kinase Assay (HEK 293) measures IGF1R receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56179 Supplier Eurofins Discoverx Supplier No. 7930505C1
The PathHunter Human IGF1R Stable Cell Line Functional Kinase Assay (HEK 293) measures IGF1R receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.