Learn More
Eurofins Discoverx PathHunter Human TrkC-P75 Functional Kinase Cell-Based Assay Kit (U2OS)
Catalog No. 5000455623
Encompass
The PathHunter Human TrkC-P75 Stable Cell Line Functional Kinase Assay (U2OS) measures TrkC-P75 receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55623 Supplier Eurofins Discoverx Supplier No. 7930531C3
The PathHunter Human TrkC-P75 Stable Cell Line Functional Kinase Assay (U2OS) measures TrkC-P75 receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.