Learn More
Eurofins Discoverx PathHunter Human TrkB-P75 Functional Kinase Cell-Based Assay Kit (U2OS)
Catalog No. 5000455724
Encompass
The PathHunter Human TrkB-P75 Stable Cell Line Functional Kinase Assay (U2OS) measures TrkB-P75 receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55724 Supplier Eurofins Discoverx Supplier No. 7930530C3
Add to Cart
Add to Cart
The PathHunter Human TrkB-P75 Stable Cell Line Functional Kinase Assay (U2OS) measures TrkB-P75 receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.