Learn More
Eurofins Discoverx PathHunter Human IκB Degradation Cell-Based Assay Kit (A549)
Catalog No. 5000455585
Encompass
The PathHunter Human IκB Degradation Stable Cell Line Assay (A549) measures pathway activation-induced degradation of IκB target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55585 Supplier Eurofins Discoverx Supplier No. 7930538C15
The PathHunter Human IκB Degradation Stable Cell Line Assay (A549) measures pathway activation-induced degradation of IκB target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.