Learn More
Eurofins Discoverx PathHunter Human IkB Degradation Cell-Based Assay Kit (THP-1)
Catalog No. 5000456221
Encompass
The PathHunter Human IkB Degradation Stable Cell Line Assay (THP-1) measures pathway activation-induced degradation of IκB target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56221 Supplier Eurofins Discoverx Supplier No. 7930538C14
The PathHunter Human IkB Degradation Stable Cell Line Assay (THP-1) measures pathway activation-induced degradation of IκB target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.