Learn More
Eurofins Discoverx PathHunter Mouse HTR2A (5-HT2A) β-Arrestin Cell-Based Assay Kit (CHO-K1)
Supplier:  Eurofins Discoverx 7930567C2
Encompass
The PathHunter Mouse HTR2A (5-HT2A) β-Arrestin Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure mHTR2A (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55937