Learn More
Eurofins Discoverx PathHunter Human AVPR1A β-Arrestin Cell-Based Assay Kit (U2OS)
Catalog No. 5000456096
Encompass
The PathHunter Human AVPR1A β-Arrestin Stable Cell Line Assay (U2OS) contains a stable clonal cell line used to measure AVPR1A (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56096 Supplier Eurofins Discoverx Supplier No. 7930587C3
The PathHunter Human AVPR1A β-Arrestin Stable Cell Line Assay (U2OS) contains a stable clonal cell line used to measure AVPR1A (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.