Learn More
Eurofins Discoverx PathHunter Mouse ADRA1B β-Arrestin Cell-Based Assay Kit (CHO-K1)
Catalog No. 5000455667
Encompass
The PathHunter Mouse ADRA1B β-Arrestin Stable Cell Pool Assay (CHO-K1) contains a stable engineered cell pool used to measure mADRA1B (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell pool vials, all required culture reagents for maintaining the cells in continuous culture, and detection reagents for running the assay.
Catalog No. 50-004-55667 Supplier Eurofins Discoverx Supplier No. 7930595P2
The PathHunter Mouse ADRA1B β-Arrestin Stable Cell Pool Assay (CHO-K1) contains a stable engineered cell pool used to measure mADRA1B (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell pool vials, all required culture reagents for maintaining the cells in continuous culture, and detection reagents for running the assay.