Learn More
Eurofins Discoverx PathHunter Mouse BRS3 β-Arrestin Cell-Based Assay Kit (U2OS)
Catalog No. 5000456114
Encompass
The PathHunter Mouse BRS3 β-Arrestin Stable Cell Pool Assay (U2OS) contains a stable engineered cell pool used to measure mBRS3 (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell pool vials, all required culture reagents for maintaining the cells in continuous culture, and detection reagents for running the assay.
Catalog No. 50-004-56114 Supplier Eurofins Discoverx Supplier No. 7930597P3
The PathHunter Mouse BRS3 β-Arrestin Stable Cell Pool Assay (U2OS) contains a stable engineered cell pool used to measure mBRS3 (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell pool vials, all required culture reagents for maintaining the cells in continuous culture, and detection reagents for running the assay.