Learn More
Eurofins Discoverx PathHunter Mouse MC3R β-Arrestin Cell-Based Assay Kit (CHO-K1)
Catalog No. 5000455770
Encompass
The PathHunter Mouse MC3R β-Arrestin Stable Cell Pool Assay (CHO-K1) contains a stable engineered cell pool used to measure mMC3R (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell pool vials, all required culture reagents for maintaining the cells in continuous culture, and detection reagents for running the assay.
Catalog No. 50-004-55770 Supplier Eurofins Discoverx Supplier No. 7930642P2
Add to Cart
Add to Cart
The PathHunter Mouse MC3R β-Arrestin Stable Cell Pool Assay (CHO-K1) contains a stable engineered cell pool used to measure mMC3R (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell pool vials, all required culture reagents for maintaining the cells in continuous culture, and detection reagents for running the assay.