Learn More
Eurofins Discoverx PathHunter Human MIP-1 alpha (CCR1) Activated GPCR Internalization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455465
Encompass
The PathHunter Human MIP-1 alpha (CCR1) Stable Cell Line Activated GPCR Internalization Assay (U2OS) contains a stable clonal cell line used to measure CCR1 (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55465 Supplier Eurofins Discoverx Supplier No. 7930651C3
The PathHunter Human MIP-1 alpha (CCR1) Stable Cell Line Activated GPCR Internalization Assay (U2OS) contains a stable clonal cell line used to measure CCR1 (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.